What Does DNA Look Like?
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S-10
EDVOS-10EA 493 DKK
EDVOS-10
What Does DNA Look Like?
Undervisningsmateriale Biologisk undervisningsmateriale Genetics Learning Activities
This lab activity shows students what real chromosomal DNA looks like and allows them to explore the procedures involved in DNA extraction.
  • For 10 lab groups
  • Complete in 30 minutes
Also required: 95% ethanol or isopropyl alcohol.
Leveringsinformation: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, minilinks, glass rod, DNA spooling rods, test tubes, and salt.