β-Cyclodextrin

Leverandør: EDQM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

B0950000
EDQMB0950000EA 0 DKK
EDQMB0950000
β-Cyclodextrin
β-Cyclodextrin
Formel: C₄₂H₇₀O₃₅
Molekylvægt: 1134.98 g/mol
MDL Number: MFCD00078139
CAS-nummer: 7585-39-9
EINECS: 231-493-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR