VWR® DNA Molecular Weight Marker, High-Range
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E255-150UGEA 9440 DKK
E255-150UG
VWR® DNA Molecular Weight Marker, High-Range
Elektroforese reagenser Molekylærvægt markører
High-Range DNA MW (molecular weight) marker offers 10 fragments ranging from 1503 to 48 502 bp. There is sufficient material for 75 to 150 assays.
  • Optimal for evaluation fragments >10 kb
  • Easy band identification