TASKI® Jontec Linosafe F1g Linoleum-Safe Stripper
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7512688
DIVY7512688EA 453 DKK
DIVY7512688
TASKI® Jontec Linosafe F1g Linoleum-Safe Stripper
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Linoleum safe stripper for removing emulsions and maintenance films from linoleum and other water-resistant hard floors. Prevents discolouring and damage to your Linoleum floor.
  • Highly effective, slightly alkaline stripper
  • Formulated for the safe removal of emulsions from linoleum floors
  • Fact acting, low foaming
  • Low odour
Benefits:
Quick and thorough removal of emulsion layers
Safe for use on all types of water-resistant hard floors, specially on Linoleum and calcium containing stones (example marble floors)
Easy and safe to use
Slightly perfumed for a pleasant use