Cleaner disinfectant, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100892312
DIVY100892312EA 1280 DKK
DIVY100892312
Cleaner disinfectant, SURE®
Renseopløsninger Disinfektanter
Concentrated cleaner disinfectant for cleaning and disinfection of all surfaces, suitable for regular use.
  • Effective in all water conditions
  • Very effective against a wide range of microorganisms, bactericidal and yeasticidal activities
  • Contains plant-based ingredients from renewable sources
  • No environmental hazard classification, no safety warnings for product in use for user
SURE® cleaner disinfectant is part of a unique range of plant-based, 100% biodegradable cleaning products that deliver professional cleaning results - while being safe for peopleMore Product Information