Autoclave Bags
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
200100 200311 200312 200310
DELT200100EA 390 DKK
DELT200100 DELT200311 DELT200312 DELT200310
Autoclave Bags
Poser Affaldsposer
Autoclave resistant bags, printed with the biohazard graphic symbol and instructions of use in five languages.
Advarsel: Do not close the bags hermetically when placing them in the autoclave at 121 °C.