Mini mailbox P650 forwarding set white
Leverandør: DAKLAPACK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
520300
DAKL520300EA 5020 DKK
DAKL520300
Mini mailbox P650 forwarding set white
Transportbeholdere
The patented MiniMailBox system is approved for use in every country as a recognised safe way of transporting biological substances category B, UN3373 / P650.
  • Flap closure
  • Made from white cardboard
Commonly used system used by companies, hospitals and testing facilities.
Leveringsinformation: The kit contains the MiniMailBox, Safetybag with bubble liner and absorbing 4 position tube holder.