Dimethylsulfoxid ≥99.5%, Reagent Grade

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

D5879-1GA D5879-100ML D5879-1L D5879-500ML D5879-25L D5879-7L D5879-2.5L D5879-230KG D5879-5L
D5879-1GAEA 3830 DKK
D5879-1GA D5879-100ML D5879-1L D5879-500ML D5879-25L D5879-7L D5879-2.5L D5879-230KG D5879-5L
Dimethylsulfoxid ≥99.5%, Reagent Grade
Dimethylsulfoxid

  • ≥95% for general laboratory use.

Formel: (CH₃)₂SO
Molekylvægt: 78.14 g/mol
Kogepunkt: 28.5 %(V)
Smeltepunkt: 18.5 °C
Massefylde: 1.104 g/cm³ (20 °C)
Flammepunkt: 87 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance complying
Identity (NMR) complying
Assay (GC) Min. 99.5 %
Water (Karl Fischer) Max. 0.1 %
Vapor Density 2.7 (vs air)
Autoignition Temperature 573 °F
Vapor Pressure 0.42 mmHg ( 20 °C)
Explosion Limit 42 %, 63 °F
Refractive Index n20/D 1.479 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR