Vertical lifeline system, Protecta® Cabloc®, 3M™
Leverandør: CAPITAL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6191035 6191032 AC320 6191036 6191033 AC325
CPSD6191035EA 0 DKK
CPSD6191035 CPSD6191032 CPSDAC320 CPSD6191036 CPSD6191033 CPSDAC325
Vertical lifeline system, Protecta® Cabloc®, 3M™
Faldbeskyttelse Faldbeskyttelsessæt
Protecta® Cabloc® Vertical Lifeline System consists of an 8 mm cable which is terminated at the top by a cable ferrule and cable clamps at the bottom.
  • Quick and easy to install
  • Easy to configure to an existing ladder and/or pylon pole
  • Versions available with or without inline shock absorber
  • Cables are available in lengths from 5 to 200 m
Top bracket anchor is either of stainless steel or galvanised steel which is connected to two rungs of the ladder for spreading the load to provide increased strength. Likewise bottom bracket is also made either ofMore Product Information