Magnetic stirring bars, with removable ring, PTFE
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FT.001.17150
COWIFT.001.17150EA 9000 DKK
COWIFT.001.17150
Magnetic stirring bars, with removable ring, PTFE
Omrørere Omrøremagneter
PTFE coated removable ring magnetic stirring bars can be used in the same way as either cylindrical or pivot ring type.