Temperature probes, platinum resistance
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
500.200.1 500.100.1 500.300.1 500.400.1 500.500.1 500.600.1 500.700.1 500.900.1 500.307.1 500.208.1 500.206.1 500.106.1
COWI500.200.1EA 1190 DKK
COWI500.200.1 COWI500.100.1 COWI500.300.1 COWI500.400.1 COWI500.500.1 COWI500.600.1 COWI500.700.1 COWI500.900.1 COWI500.307.1 COWI500.208.1 COWI500.206.1 COWI500.106.1
Temperature probes, platinum resistance
Temperatur prober
Platinum resistance temperature probes made of stainless steel, encapsulated in PTFE, with push-on ferrule.
  • Inert and non contaminating
  • Pressure resistent
  • Features 1 m long PFA insulated cable
Response: 50%, 6 to 14 seconds
Leveringsinformation: Supplied without plug.