Magnetic stirring bar retrievers, flexible, PTFE
Leverandør: COWIE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
004.330F
COWI004.330FEA 811 DKK
COWI004.330F
Magnetic stirring bar retrievers, flexible, PTFE
Omrørere Omrøreudstyr
Retrivers are used for the removal of stirrer magnets from vessels of all kinds.