Büchner funnels, PYREX®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36060-30C
CORN36060-30CEA 0 DKK
CORN36060-30C
Büchner funnels, PYREX®
Tragte Filtreringstragte Büchner-tragte
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
Advarsel: Disc should not be subjected to pressure differential in excess of 1,01 bar.