Laminar flow cabinets, CrossFlow CLF series

Leverandør: CleanAir by Baker
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

1560401 1560402 1560403 1560404 1560405 1560406
CLEA1560401EA 70840 DKK
CLEA1560401 CLEA1560402 CLEA1560403 CLEA1560404 CLEA1560405 CLEA1560406
Laminar flow cabinets, CrossFlow CLF series
Stinkskabe og kabinetter Laminar flow-bænke
The CrossFlow series is a high quality crossflow cabinet range for handling non-hazardous materials that require a clean working environment. The ergonomically designed HEPA filtered horizontal laminar airflow (crossflow) creates an ISO Class 5 (14644-1) / GMP Class A (Annex 1) work area with low noise level and prevents ambient air from entering the work area, ensuring product protection.

  • Microprocessor flow control
  • User-friendly control panel for intuitively operating
  • HEPA filter H14 (efficiency of 99,995% MPPS) and pre-filter (ISO 16890)
  • Energy saving
More Product Information
Bestil nu

Specifikationer

Model CLF 475 CLF 575 CLF 675
Environmentally Preferable
Lighting >1000 lux >1000 lux >1000 lux
Støjniveau <58 dB(A) <58 dB(A) <54 dB(A)
Effekt 151 kW 198 kW 245 kW
Filtre HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
Electrical supply 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz
Ydermål B×D×H 1270×893×1153 mm 1574×893×1153 mm 1880×893×1153 mm
Int. W×D×H 1249×570×736 mm 1554×570×736 mm 1859×570×736 mm
Weight 110 kg 150 kg 190 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR