Face shields, ArcPro™ anti electric arc systems
Leverandør: CENTURION SAFETY PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S25WCONTAEA S25WCEAEA S506S57 S512S57 S910AEA S25WR820
CENUS25WCONTAEAEA 790 DKK
CENUS25WCONTAEA CENUS25WCEAEA CENUS506S57 CENUS512S57 CENUS910AEA CENUS25WR820
Face shields, ArcPro™ anti electric arc systems
Ansigtsskærm
These systems have been designed to protect against arc flash risks posed by electrical faults.
  • Clear PC screen protects against short circuit arcs
  • Green PC screens also feature anti-fog coating
  • Face screens supplied with chin guard made of fibre reinforced nylon
The face screens are available without or in complete kits with helmet.
Certificeringer: EN 166 1 B 8 9 (EN 170 2-1,2); Arc exposure test according to: GS-ET-29:2010, Class 2 (423 kJ/m²)