Gas Chromatograph with FID (GC-FID)
Leverandør: LUCIDITY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
928270-240
CCDL928270-240EA 135030 DKK
CCDL928270-240
Gas Chromatograph with FID (GC-FID)
Kromatografisystemer Gaskromatografer
Compact, economical, fully functional gas chromatograph.
  • Lucidity quick column change for quick and easy column installation
  • Equipped with a split/splitless injection port
  • Contains a very small oven which allows for rapid heating and cooling
  • Intuitive Lucidity Software included with purchase of instrument
  • Instrument changes color to indicate status
  • Self-installable and ease of use design
  • Low power consumption
The Lucidity GC has one of the smallest footprints of any fully functional GC-FID. Changing GC columns in the Lucidity GC is as simple asMore Product Information