Methyl arachidonate

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

90014-100 90014-25 90014-50
CAYM90014-100EA 2700 DKK
CAYM90014-100 CAYM90014-25 CAYM90014-50
Methyl arachidonate
Methyl arachidonate
Formel: C₂₁H₃₄O₂
MDL Number: MFCD00016775
CAS-nummer: 2566-89-4
EINECS: 219-900-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR