2'3'-cGAMP ELISA Kit
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
501700-96
CAYM501700-96EA 9570 DKK
CAYM501700-96
2'3'-cGAMP ELISA Kit
Assays ELISA'er
For the quantification of 2’3’-cGAMP in cell lysates.
  • Measure 2'3'-cGAMP in cell lysates
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Lower limit of detection (LLOD) is 9.6 pg/ml (0.01 pmol/ml)
  • Run overnight or incubate for just 2 hours without compromising sensitivity
  • Monitor the kinetics of 2'3'-cGAMP formation and hydrolysis in a biological setting
  • Identify compounds that modulate cGAS activity
Cayman’s 2’3’-cGAMP ELISA Kit is a competitive assay that can be used for quantification of 2’3’-cGAMP in cell lysates. This assay has a range from 100 ng/ml to 6,1 pg/ml with a midpoint of approximately 900 pg/ml (50% B/B0) and a sensitivity of approximately 85 pg/ml (80% B/B0). Powered by BIOLOG Life Science Institute.