(±)-11-HDoHE

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

33450-25 33450-50 33450-100 33450-250
CAYM33450-25EA 1630 DKK
CAYM33450-25 CAYM33450-50 CAYM33450-100 CAYM33450-250
(±)-11-HDoHE
(±)-11-HDoHE
Formel: C₂₂H₃₂O₃
CAS-nummer: 87018-59-5
UN nummer: 1170
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR