(±)-8-HDoHE

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

33350-25 33350-50 33350-100 33350-250
CAYM33350-25EA 1630 DKK
CAYM33350-25 CAYM33350-50 CAYM33350-100 CAYM33350-250
(±)-8-HDoHE
(±)-8-HDoHE
Formel: C₂₂H₃₂O₃
CAS-nummer: 90780-54-4
UN nummer: 1170
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR