Anti-COX-1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CX111]

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

160120-1
CAYM160120-1EA 7360 DKK
CAYM160120-1
Anti-COX-1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CX111]
Antistoffer
Type: Primary
Antigen: COX-1
Clonality: Monoclonal
Clone: CX111
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Mouse, Sheep, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR