Lansoprazole

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

13627-500 13627-1 13627-5 13627-10
CAYM13627-500EA 820 DKK
CAYM13627-500 CAYM13627-1 CAYM13627-5 CAYM13627-10
Lansoprazole
Lansoprazole
Formel: C₁₆H₁₄F₃N₃O₂S
Molekylvægt: 369.36 g/mol
MDL Number: MFCD00866873
CAS-nummer: 103577-45-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR