Anti-RAF-1 Rabbit Polyclonal Antibody

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

10009504-1
CAYM10009504-1EA 8800 DKK
CAYM10009504-1
Anti-RAF-1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RAF-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR