Leukotriene B₄ Express ELISA kit
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10009292-96S 10009292-96 10009292-480S 10009292-480
CAYM10009292-96SEA 7160 DKK
CAYM10009292-96S CAYM10009292-96 CAYM10009292-480S CAYM10009292-480
Leukotriene B₄ Express ELISA kit
Assays ELISA'er
A competitive assay that can be used for quantification of LTB₄ in plasma and other sample matrices.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTB₄ levels down to 45 pg/ml
  • Rapid assay; get results in under five hours
LTB₄ stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB₄ is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB₄ and then further oxidized to ω-carboxy dinor LTB₄ and ω-carboxy tetranor-LTB3. It is not excreted in the urine.
Leveringsinformation: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.