Genistein

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

10005167-100 10005167-250 10005167-500 10005167-1
CAYM10005167-100EA 662 DKK
CAYM10005167-100 CAYM10005167-250 CAYM10005167-500 CAYM10005167-1
Genistein
Genistein
Formel: C₁₅H₁₀O₅
Smeltepunkt: 297…298 °C
MDL Number: MFCD00016952
CAS-nummer: 446-72-0
EINECS: 207-174-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR