L (+)-Glutaminsyre

Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

DLM-3725-0.5
CAMBDLM-3725-0.5EA 0 DKK
CAMBDLM-3725-0.5
L (+)-Glutaminsyre
L (+)-Glutaminsyre
Formel: HO₂CCH₂CH₂CH(NH₂)CO₂H
Kogepunkt: 325 °C (1013 hPa)
Smeltepunkt: 160 °C
Massefylde: 1,538 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002634
CAS-nummer: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR