Anti-EF-Tu Rabbit Polyclonal Antibody [clone: L338]

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS9146
BWRLBS9146EA 2150 DKK
BWRLBS9146
Anti-EF-Tu Rabbit Polyclonal Antibody [clone: L338]
Antistoffer
Synthetic peptide, corresponding to amino acids 300-350 of Human EF-Tu.

EF-Tu (L338) pAb detects endogenous levels of EF-Tu protein.

Type: Primary
Antigen: EF-Tu
Clonality: Polyclonal
Clone: L338
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR