Anti-SHP-1 Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS6827
BWRLBS6827EA 2800 DKK
BWRLBS6827
Anti-SHP-1 Rabbit Polyclonal Antibody
Antistoffer
Synthetic peptide, corresponding to amino acids 550-600 of Human EphA2.

SHP-1 pAb detects endogenous levels of SHP-1 protein.

Type: Primary
Antigen: SHP-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR