Anti-BDNF Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS6533
BWRLBS6533EA 2800 DKK
BWRLBS6533
Anti-BDNF Rabbit Polyclonal Antibody
Antistoffer
Recombinant full length Human BDNF.

BDNF pAb detects endogenous levels of BDNF protein.

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR