Anti-CD133 Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS6480
BWRLBS6480EA 2800 DKK
BWRLBS6480
Anti-CD133 Rabbit Polyclonal Antibody
Antistoffer
Recombinant full length Human CD133.

CD133 pAb detects endogenous levels of CD133 protein.

Type: Primary
Antigen: CD133
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR