Anti-CtBP2 Rabbit Polyclonal Antibody [clone: P432]

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS2287
BWRLBS2287EA 2150 DKK
BWRLBS2287
Anti-CtBP2 Rabbit Polyclonal Antibody [clone: P432]
Antistoffer
Synthetic peptide, corresponding to amino acids 400-450 of Human CtBP2.

CtBP2 (P432) pAb detects endogenous levels of CtBP2 protein.

Type: Primary
Antigen: CtBP2
Clonality: Polyclonal
Clone: P432
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR