Anti-MST1/2 Rabbit Polyclonal Antibody [clone: M178]

Leverandør: Bioworld Technology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS1554
BWRLBS1554EA 2150 DKK
BWRLBS1554
Anti-MST1/2 Rabbit Polyclonal Antibody [clone: M178]
Antistoffer
Synthetic peptide, corresponding to amino acids 150-200 of Human MST1.

MST1/2 (M178) pAb detects endogenous levels of MST1/2 protein.

Type: Primary
Antigen: MST1/2
Clonality: Polyclonal
Clone: M178
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR