Reducing tubing connectors, straight
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8703-0408 8703-0412 8703-0812
BURK8703-0408EA 309 DKK
BURK8703-0408 BURK8703-0412 BURK8703-0812
Reducing tubing connectors, straight
Fittings og stik
Made from PP, white.
  • Sterilisable
  • Autoclavable up to max. 121 °C (20 min.)
  • Good chemical resistance
Reducing nozzles for the reduction from bigger to small tubings.