Human recombinant TrkA-Fc Chimera (from HEK293 cells)
Leverandør: Biosensis
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PE-1248-100 PE-1248-50
BSENPE-1248-100EA 2690 DKK
BSENPE-1248-100 BSENPE-1248-50
Human recombinant TrkA-Fc Chimera (from HEK293 cells)
Proteiner
TrkA is a member of the neurotrophic tyrosine kinase receptor family. It is a membrane-bound receptor that upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. TrkA is required for high-affinity binding to nerve growth factor (NGF), neurotrophin-3 and neurotrophin-4/5 but not brain-derived neurotrophic factor (BDNF). TrkA leads to cell differentiations and may play a role in specifying sensory neuron subtypes. It has a crucial role in the development and function of the nociceptive reception system as well as establishment of thermal regulation via sweating. SUBUNIT: Exists in a dynamic equilibrium between monomeric (low affinity) and dimeric (high affinity) structures. SUBCELLULAR LOCATION: TrkA is a heavily glycosylated transmembrane protein and contains 13 potential N-linked glycosylation sites. The TrkA-Fc chimera has N-linked and may have O-linked oligosaccharides.