Anti-GST Mouse Monoclonal Antibody [clone: BGSTB6]

Leverandør: Biosensis
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M-1306-100
BSENM-1306-100EA 2690 DKK
BSENM-1306-100
Anti-GST Mouse Monoclonal Antibody [clone: BGSTB6]
Antistoffer
GST (Glutathione S-Transferase) is a 26 kDa protein encoded by the Schistosoma japonicum.

Mouse monoclonal anti-GST tag antibody detects over expressed glutathione- transferase (GST) fusion proteins.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH) and Immunoprecipitation (IP). Suggested dilutions for WB of 1:1000, IS and IP at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: GST
Clonality: Monoclonal
Clone: BGSTB6
Conjugation: Unconjugated
Epitope: GST tag
Host: Mouse
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR