Multi-Neurotrophin Rapid™ screening ELISA kit: Human
Leverandør: Biosensis
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BEK-2227
BSENBEK-2227EA 4390 DKK
BSENBEK-2227
Multi-Neurotrophin Rapid™ screening ELISA kit: Human
Assays ELISA'er
The Biosensis Multi-Neurotrophin Rapid™ Screening ELISA Kit has been designed to allow rapid screening and quantification of NGF, BDNF, NT3 and NT4/5 in cell culture supernatants, lysates, serum, plasma (EDTA and citrate) and brain extracts only if used as directed.
This two-plate kit consists of four sets of 6 strips for each Neurotrophin, allowing for the assay of 16 samples per Neurotrophin tested and a full range of standards, all run in duplicate. It provides the identical sensitivities and ranges that are achieved in the complete, individual ELISA kits, thus allowing easy progression into the larger individual assay sizes when needed.
The Multi-Neurotrophin Rapid™ Screening ELISA Kit presents a cost effective way to quickly screen multiple samples, which can then be published or used prior to a more extensive analysis with the individual kits. The kit comes complete with all detection reagents and is ready to use. Individual coated strips are provided and each set of standards and detection antibodies are colour-coded.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular human serum and plasma samples.
Leveringsinformation: Each kit contains 2 × 96 pre-coated strip plates (6 strips/48 wells per neurotrophin target), protein standards, detection reagents, wash and sample buffers and detailed protocols.
Advarsel: For research use only. Not to be used in diagnostic or clinical procedures.