Paste, silicone
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
61605
BRND61605EA 394 DKK
BRND61605
Paste, silicone
Smøremidler
For greasing ground joints.
  • Low viscosity
  • Operating temperature –40 to +160 °C