Compact automatic burettes, Schellbach stripe, SILBERBRAND
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23809 23810 23811
BRND23809EA 1330 DKK
BRND23809 BRND23810 BRND23811
Compact automatic burettes, Schellbach stripe, SILBERBRAND
Buretter
AR® glass, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, reduced distance between subdivision marks
  • Detachable PTFE stopcock
Error limits according to class B, DIN EN ISO 385.
Bestillingsoplysninger: Please order supports and burette clamps separately.
Leveringsinformation: Supplied as burette tube, stopcock with precision tip, filling tube (PVC) and 1000 ml PE bottle with base.