Oil drum top covers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
813784
BRDY813784EA 1130 DKK
BRDY813784
Oil drum top covers
Spild materialer
Oil sorbent drum top cover with pre-cut holes that absorbs oil and petroleum based fluids.
This helps to keep drum top, work floor and surrounding storage area neat and clean by preventing drips from reaching the floor.