Label Printer, AZERTY with Bluetooth and Workstation PWID Suite, M610
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
317800
BRDY317800EA 7280 DKK
BRDY317800
Label Printer, AZERTY with Bluetooth and Workstation PWID Suite, M610
Printere Label Printers
Get the durability your work demands.
  • Design and print labels using a keypad, mobile app or desktop software
  • Work longer with an enhanced Gen3 Li-ION battery – print 4500 labels on a full charge
  • Stay connected wherever your work happens with enhanced Bluetooth technology
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 50,80 mm per second print speed
  • Get added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 1,80 m drops