Reversible temperature indicator labels
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
195844
BRDY195844EA 165 DKK
BRDY195844
Reversible temperature indicator labels
Temperaturindikatorstrips
These reversible temperature indicator labels designed to display temperatures of food and medical supply packaging or of components during research, repair and maintenance.
  • Changes colour at designated temperatures
  • Temperature ranges between 0 to 100 °C
  • Highly customisable
  • Permanent acrylic adhesive
Colour: Red and black.