Printer Ribbons, M710 R6400 Series
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
173189
BRDY173189EA 604 DKK
BRDY173189
Printer Ribbons, M710 R6400 Series
Printer bånd
One-color ribbon for printing a single color onto a label.
  • R6400 series resin formulation offers superior chemical and abrasion resistance when printed on the appropriate Brady® label material
  • Recommended for use with materials designed to resist chemicals, such as B-492 frozen surface polyester and B-481 chemical resistant polyester
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and theMore Product Information