Labeling tapes, floor marking arrows, B-514, ToughStripe™
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
104554
BRDY104554EA 1860 DKK
BRDY104554
Labeling tapes, floor marking arrows, B-514, ToughStripe™
Tape
Extremely durable ToughStripe™ die-cut floor marking tapes are made of rugged polyester with an ultra-aggressive adhesive that will hold up against forklift traffic.
  • Brady B-514 is a surface printed polyester with a rigid, clear polyester overlaminate
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands heavy traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Permanent rubber