Anti-NDE1 Rabbit Polyclonal Antibody

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-9316R
BOSSBS-9316REA 5410 DKK
BOSSBS-9316R
Anti-NDE1 Rabbit Polyclonal Antibody
Antistoffer
Required for centrosome duplication. Essential for the development of the cerebral cortex. May regulate the production of neurons by controlling the orientation of the mitotic spindle during division of cortical neuronal progenitors of the proliferative ventricular zone of the brain. Orientation of the division plane perpendicular to the layers of the cortex gives rise to two proliferative neuronal progenitors whereas parallel orientation of the division plane yields one proliferative neuronal progenitor and a post-mitotic neuron. A premature shift towards a neuronal fate within theMore Product Information
Bestil nu

Specifikationer

Antigen symbol NDE1
Environmentally Preferable
Antigen name NudE Nuclear Distribution Gene E homolog 1
Antigen synonymer NDE|LIS4|MHAC|HOM-TES-87|NUDE|NUDE1
Antistof type Primary
Clonality Polyclonal
Konjugation Unconjugated
Reactivity Human,Rat,Mouse
Vært Rabbit
Gen ID 54820
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A.
Koncentration 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR