Anti-RENBP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-8497R-A750
BOSSBS-8497R-A750EA 6670 DKK
BOSSBS-8497R-A750
Anti-RENBP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
RENBP is a 427 amino acid protein that is a proteinaceous renin inhibitor. In inhibiting renin, RENBP forms a complex with it; a high molecular weight renin. RENBP contains a leucine zipper domain, which is essential for its dimerisation with renin. RENBP can catalyze the interconversion of N-acetylglucosamine to N-acetylmannosamine, indicating that it is a GlcNAc 2-epimerase. Sequences of porcine, human and rat renin-binding proteins are highly homologous. The RENBP gene is conserved in dog, mouse, rat and zebrafish, and maps to human chromosome Xq28 between DXS52 and G6PD. Rat Renbp isMore Product Information