Anti-PANK4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-8340R-A647
BOSSBS-8340R-A647EA 6670 DKK
BOSSBS-8340R-A647
Anti-PANK4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
PANK4 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the physiological regulation of the intracellular coenzyme A (CoA) concentration. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PANK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR