Anti-ASNA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-7972R-A350
BOSSBS-7972R-A350EA 6670 DKK
BOSSBS-7972R-A350
Anti-ASNA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
ASNA1 is the human homolog of the E.coli arsA gene which is an ATPase, and is the catalytic component of a multisubunit oxyanion pump responsible for resistance to arsenicals and antimonials.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR