Anti-CPT1B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BS-5045R-FITC
BOSSBS-5045R-FITCEA 6880 DKK
BOSSBS-5045R-FITC
Anti-CPT1B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene. [provided by RefSeq, Jun 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Bestil nu

Specifikationer

Antigen symbol CPT1B
Antigen name Carnitine Palmitoyltransferase 1B
Environmentally Preferable
Antigen synonymer CPT1B|FLJ58750|carnitine palmitoyltransferase 1B (muscle)|carnitine palmitoyltransferase 1B (muscle) |KIAA1670|CPTI|FLJ55729|carnitine palmitoyltransferase 1A (muscle)|carnitine O-palmitoyltransferase I|carnitine palmitoyltransferase I|M-CPT1|muscle|mitochondrial muscle isoform|carnitine palmitoyltransferase I-like|CPT1-M|carnitine palmitoyltransferase 1B|MCCPT1|CPT1M|MCPT1
Antistof type Primary
Clonality Polyclonal
Konjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Vært Rabbit
Gen ID 1375
Isotype IgG
ImmunoChemistry Yes
Immunogen 530-580/772
Form liquid
Purification Purified by Protein A
Koncentration 1 μg/μl
Krydsadsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opbevaringstemperatur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR