Anti-CPT1B Rabbit Polyclonal Antibody (Cy3®)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-5045R-CY3
BOSSBS-5045R-CY3EA 6880 DKK
BOSSBS-5045R-CY3
Anti-CPT1B Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene. [provided by RefSeq, Jun 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200