Anti-BMP15 Rabbit Polyclonal Antibody
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-4720R
BOSSBS-4720REA 5410 DKK
BOSSBS-4720R
Anti-BMP15 Rabbit Polyclonal Antibody
Antistoffer
Required for ovarian folliculogenesis. Promotes primordial follicle development. Stimulates granulosa cell proliferation. Promotes cell transition from G0/G1 to S and G2/M phases, through an increase of CCND1 and CCNE1 expression, and RB1 phosphorylation. It regulates STAR expression and cAMP-dependent progesterone release in granulosa and thecal cells. Attenuates the suppressive effects of activin A on STAR expression and progesterone production by increasing the expression of inhibin B. It suppresses FST and FSTL3 production in granulosa-lutein cells.
WB(1:100-1000), IHC-P(1:100-500),More Product Information